Anti-EIF3E Rabbit Polyclonal Antibody
Catalog # ABNOPAB10203
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Eukaryotic translation initiation factor 3, subunit E
- Antigen symbol:EIF3E
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Monkey,Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:16341
- Antigen synonyms:EIF3S6|eIF3-p46|subunit 6 48kDa|eukaryotic translation initiation factor 3|murine mammary tumor integration site 6 (oncogene homolog)|EIF3-P48|subunit 6 (48kD)|mammary tumor-associated protein INT6|INT6
- Storage buffer:20 mM KH2PO4, 150 mM NaCl, pH 7,2 (0,01% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to C-terminus of mouse Eif3e.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of Eif3e.
Recommended Dilutions: ELISA: 1:90000; Western Blot: 1:1000
Type: Primary
Antigen: EIF3E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Dog, Human, Monkey, Mouse, Rat