Anti-HSP90AA1 Rabbit Polyclonal Antibody
ABNOPAB10219
: Abnova
- Antibody type:Primary
- Antigen name:Heat shock protein 90kDa alpha (cytosolic), class A member 1
- Antigen symbol:HSP90AA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Drosophila,Monkey,Chicken,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:3320
- Antigen synonyms:HSPCA|Hsp89|EL52|HSP86|LAP-2|HSPCAL1|HSPN|Hsp90|HSPC1|HSP90N|HSPCAL4|LAP2|HSP89A|HSP90A
- Storage buffer:20 mM KH2PO4, 150 mM NaCl, pH 7,2 (0,01% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to residues surrounding K294 of human HSP90AA1.
- Size:100 μg
- Pk:100 µl
Rabbit polyclonal antibody raised against synthetic peptide of HSP90AA1.
Recommended Dilutions: ELISA: 1:2500-1:10000; Western Blot: 1:500-1:2500; Immunohistochemistry: 5-10 ?g/ml
Type: Primary
Antigen: HSP90AA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken, Drosophila, Human, Monkey, Mouse, Rat