Anti-TLR11 Rabbit Polyclonal Antibody
Catalog # ABNOPAB0229
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Toll-like Receptor 11
- Antigen symbol:TLR11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- Reactivity:Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:239081
- Antigen synonyms:Gm287|MGC129353|MGC129352
- Amino acid number:900 to 950
- Storage buffer:PBS (0,05% BSA, 0,05% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 900-950 of mouse Tlr11.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of Tlr11.
Type: Primary
Antigen: TLR11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse