Order Entry
Export
ContactUsLinkComponent
 
Anti-ITGA5 Rat Monoclonal Antibody [clone: 5H10-27]
Catalog # ABNOMAB3234
Supplier:  Abnova
Anti-ITGA5 Rat Monoclonal Antibody [clone: 5H10-27]
Catalog # ABNOMAB3234
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Integrin Alpha 5
  • Antigen symbol:
    ITGA5
  • Clonality:
    Monoclonal
  • Clone:
    5H10-27
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Mouse
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    16402
  • Antigen synonyms:
    FNRA|CD49e|Fibronectin Receptor Alpha Polypeptide|VLA-5|VLA5A
  • Storage buffer:
    100 mM BBS, pH 8,2
  • Storage temperature:
    Store at 4 °C.
  • Immunogen:
    Native Itga5 from mouse mast cell line MC/9.
  • Size:
    500 μg
  • Pk:
    500 µg

Specifications

About this item

Rat monoclonal antibody raised against Itga5.

Recommended Dilutions: Flow Cytometry: 1 ?g/10^6 cells

Type: Primary
Antigen: ITGA5
Clonality: Monoclonal
Clone: 5H10-27
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse