Order Entry
Export
ContactUsLinkComponent
Anti-Luciferase Mouse Monoclonal Antibody [clone: Luci 21 1-107]
Anti-Luciferase Mouse Monoclonal Antibody [clone: Luci 21 1-107]
Catalog # ABNOMAB2322
Supplier:  Abnova
Anti-Luciferase Mouse Monoclonal Antibody [clone: Luci 21 1-107]
Catalog # ABNOMAB2322
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Luciferase
  • Clonality:
    Monoclonal
  • Clone:
    Luci 21 1-107
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Firefly
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Storage buffer:
    buffer containing 0,1% sodium azide
  • Storage temperature:
    Store at –20 °C or –80 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified luciferase protein from firefly, Photinus pyralis.
  • Size:
    100 μl
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against native luciferase.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: Luciferase
Clonality: Monoclonal
Clone: Luci 21 1-107
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Firefly