Order Entry
Export
ContactUsLinkComponent
Anti-TRA-1-60 Mouse Monoclonal Antibody [clone: TRA-1-60(R)]
Anti-TRA-1-60 Mouse Monoclonal Antibody [clone: TRA-1-60(R)]
  ABNOMAB2391
 :  Abnova
Anti-TRA-1-60 Mouse Monoclonal Antibody [clone: TRA-1-60(R)]
  ABNOMAB2391
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Podocalyxin-like 1
  • Antigen symbol:
    PODXL1
  • Clonality:
    Monoclonal
  • Clone:
    TRA-1-60(R)
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgM
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Antigen synonyms:
    PCLP|PCLP-1|keratan sulfate antigen TRA1-60|Gp200|PC
  • Storage buffer:
    buffer containing 0,1% sodium azide
  • Storage temperature:
    Store at 4 °C for short term. For long term storage store at –20 °C or –80 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified from human embryonal carcinoma cell line 2102Ep.
  • Size:
    100 μl
  • Pk:
    100 µg

 

 

Mouse monoclonal antibody raised against native TRA-1-60.

Recommended Dilutions: Flow Cytometry: 1:50-1:200; Immunocytochemistry: 1:50-1:200; Immunofluorescence: 1:50-1:200

Type: Primary
Antigen: TRA-1-60
Clonality: Monoclonal
Clone: TRA-1-60(R)
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human