Anti-TERF1 Mouse Monoclonal Antibody [clone: Jun-57]
Catalog # ABNOMAB2400
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Telomeric Repeat Binding Factor 1
- Antigen symbol:TERF1
- Clonality:Monoclonal
- Clone:Jun-57
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Primates,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:7013
- Antigen synonyms:TRF1|hTRF1-AS|TRBF1|PIN2|t-TRF1|TRF
- Storage buffer:Tris-glycine, 150 mM NaCl (0,05% sodium azide)
- Storage temperature:Store at 4 °C for short term. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human TERF1.
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against recombinant TERF1.
Recommended Dilutions: Immunocytochemistry: 1:50-1:200; Immunofluorescence: 1:50-1:200; Western Blot: 0.5 ?g/ml
Type: Primary
Antigen: TERF1
Clonality: Monoclonal
Clone: Jun-57
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Primates, Rat