Order Entry
Export
ContactUsLinkComponent
Anti-BFSP1 Mouse Monoclonal Antibody [clone: FIL-27]
Anti-BFSP1 Mouse Monoclonal Antibody [clone: FIL-27]
Catalog # ABNOMAB2423
Supplier:  Abnova
Anti-BFSP1 Mouse Monoclonal Antibody [clone: FIL-27]
Catalog # ABNOMAB2423
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    BFSP1
  • Clonality:
    Monoclonal
  • Clone:
    FIL-27
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Bovine
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Lyophilized
  • Gene ID:
    631|281644
  • Storage buffer:
    1.2% sodium acetate (2 mg BSA, 0,01 mg sodium azide)
  • Storage temperature:
    Store at –20 °C on dry atmosphere. After reconstitution with 1 mL of 1.2% sodium acetate or neutral PBS and concentration will be 100 ug/mL, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native from human and bovine lens filament enriched fraction (plasma membrane-cytoskeleton complex).
  • Purification:
    Affinity purification
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against native BFSP1.

Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunohistochemistry (Frozen sections): 2-4 ?g/ml

Type: Primary
Antigen: BFSP1
Clonality: Monoclonal
Clone: FIL-27
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human