Order Entry
Export
ContactUsLinkComponent
Anti-DHX58 Rabbit Polyclonal Antibody
Anti-DHX58 Rabbit Polyclonal Antibody
Catalog # ABNOPAB13438
Supplier:  Abnova
Anti-DHX58 Rabbit Polyclonal Antibody
Catalog # ABNOPAB13438
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    DEXH (Asp-Glu-X-His) box polypeptide 58
  • Antigen symbol:
    DHX58
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    79132
  • Antigen synonyms:
    RLR-3|D11LGP2|D11lgp2e|Probable ATP-dependent RNA helicase DHX58|LGP2|D11LGP2E|Probable ATP-dependent helicase LGP2
  • Storage buffer:
    PBS (0,02% sodium azide)
  • Storage temperature:
    Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to internal region 11 amino acids of human DHX58.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of DHX58.

Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: DHX58
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat