Anti-PROCR Rat Monoclonal Antibody [clone: LMR-42]
Catalog # ABNOMAB3398
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Protein C receptor, endothelial
- Antigen symbol:PROCR
- Clonality:Monoclonal
- Clone:LMR-42
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Form:Culture Supernatant
- Gene ID:10544
- Antigen synonyms:EPCR|CCCA|CCD41
- Storage buffer:0,1% sodium azide
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length human PROCR.
- Size:1 mL
- Pk:1 ml
Specifications
About this item
Rat monoclonal antibody raised against full length recombinant PROCR.
Recommended Dilutions: Immunohistochemistry (Frozen sections): 1:20
Type: Primary
Antigen: PROCR
Clonality: Monoclonal
Clone: LMR-42
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human