Anti-H5 Mouse Monoclonal Antibody [clone: B200M]
ABNOMAB4826
: Abnova
- Antibody type:Primary
- Antigen name:Influenza A virus Haemagglutinin H5 (Avian)
- Antigen symbol:H5
- Clonality:Monoclonal
- Clone:B200M
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Virus
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS (0,02% sodium azide)
- Storage temperature:Store at 4 °C.
- Immunogen:A synthetic peptide corresponding to 14 amino acids near internal region of IInfluenza A Virus Haemagglutinin H5 (Avian) Hemagglutinin.
- Pk:100 µG
Mouse monoclonal antibody raised against synthetic peptide of Influenza A Virus Haemagglutinin H5.
Recommended Dilutions: ELISA: detect 10ng of free peptide at 1 ?g/ml
Type: Primary
Antigen: H5
Clonality: Monoclonal
Clone: B200M
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Virus