Anti-CD59 Monoclonal Antibody (Biotin) [clone: MEM-43]
Catalog # ABNOMAB5009
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:CD59 Molecule, Complement Regulatory Protein
- Antigen symbol:CD59
- Clonality:Monoclonal
- Clone:MEM-43
- Conjugation:Biotin
- Flow cytometry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:966
- Antigen synonyms:MIRL|HRF-20|MEM43|1F5|MACIF|MIN3|EJ16|MSK21|EL32|HRF20|p18-20|MIC11|MIN1|16.3A5|MIN2|MAC-IP|G344|EJ30
- Storage buffer:PBS, pH 7,4 (15 mM sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified CD59 from thymocytes and T lymphocytes.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against native CD59.
Recommended Dilutions: Flow Cytometry: 1:40
Type: Primary
Antigen: CD59
Clonality: Monoclonal
Clone: MEM-43
Conjugation: Biotin
Epitope:
Host:
Isotype: IgG2a
Reactivity: Human