Anti-CD22 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-01]
Catalog # ABNOMAB5034
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:CD22 molecule
- Antigen symbol:CD22
- Clonality:Monoclonal
- Clone:MEM-01
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Primates,Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:933
- Antigen synonyms:SIGLEC-2|SIGLEC2
- Storage buffer:PBS (0,2% BSA, 15 mM sodium azide)
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified CD22 from Raji Burkitt's lymphoma cell line.
- Size:100 Reactions
- Pk:100 Tests
Specifications
About this item
Mouse monoclonal antibody raised against native CD22.
Recommended Dilutions: Flow Cytometry: 20 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)
Type: Primary
Antigen: CD22
Clonality: PE (Phycoerythrin)
Clone: Monoclonal
Conjugation: MEM-01
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: Human, Primates