Anti-CD14 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-15]
Catalog # ABNOMAB5064
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:CD14 Molecule
- Antigen symbol:CD14
- Clonality:Monoclonal
- Clone:MEM-15
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Primates,Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:929
- Antigen synonyms:monocyte differentiation antigen CD14|myeloid cell-specific leucine-rich glycoprotein
- Storage buffer:PBS (0,2% BSA, 15 mM sodium azide)
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified human CD14.
- Size:100 Reactions
- Pk:100 Tests
Specifications
About this item
Mouse monoclonal antibody raised against native CD14.
Recommended Dilutions: Flow Cytometry: 20 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)
Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: MEM-15
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Primates