Anti-DDX58 Rabbit Polyclonal Antibody
Catalog # ABNOPAB12973
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:DEAD (Asp-Glu-Ala-Asp) box polypeptide 58
- Antigen symbol:DDX58
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:23586
- Antigen synonyms:Probable ATP-dependent RNA helicase DDX58|DEAD box protein 58|RIGI|RIG-I|RIG-1|RLR-1
- Storage buffer:PBS (0,02% sodium azide)
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant GST fusion protein corresponding to human DDX58.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit Polyclonal antibody raised against recombinant DDX58.
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: DDX58
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat