Anti-Neurofilament heavy Protein Mouse Monoclonal Antibody [clone: NF-01]
Catalog # ABNOMAB3638
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Neurofilament heavy Protein
- Clonality:Monoclonal
- Clone:NF-01
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Pig,Mammals
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS, pH 7,4 (15 mM sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified porcine Neurofilament heavy protein.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against native Neurofilament heavy protein .
Type: Primary
Antigen: Neurofilament heavy Protein
Clonality: Monoclonal
Clone: NF-01
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mammals, Pig