Anti-CADM1 Rabbit Polyclonal Antibody
ABNOPAB12107
: Abnova
- Antibody type:Primary
- Antigen name:Cell adhesion molecule 1
- Antigen symbol:CADM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Form:Antiserum
- Gene ID:23705
- Antigen synonyms:synCAM1|IGSF4|sgIGSF|SYNCAM|BL2|IGSF4A|TSLC1|NECL2|ST17|Necl-2|sTSLC-1|RA175
- Storage buffer:0,02% sodium azide
- Storage temperature:Store at 4 °C for short term. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to C-terminus of rat CADM1.
- Size:100 μl
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic peptide of CADM1.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: CADM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat