Anti-IMP38 Mouse Monoclonal Antibody [clone: IPO-38]
ABNOMAB2092
: Abnova
- Antibody type:Primary
- Antigen name:Importin 38
- Antigen symbol:IMP38
- Clonality:Monoclonal
- Clone:IPO-38
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgM kappa
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Antigen synonyms:MGC117425|FLJ31471|MGC104230|HMG14|FLJ27265
- Storage buffer:20 mM KH2PO4, 500 mM NaCl, pH 7,2 (0,01% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified from spleen cells of a human patient with hairy cell leukemia.
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against native IPO-38.
Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:50-1:100
Type: Primary
Antigen: IMP38
Clonality: Monoclonal
Clone: IPO-38
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human, Mouse, Rat