Anti-TRPM7 Rabbit Polyclonal Antibody
ABNOPAB12118
: Abnova
- Antibody type:Primary
- Antigen name:Transient Receptor Potential Cation Channel Subfamily M member 7
- Antigen symbol:TRPM7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Form:Antiserum
- Gene ID:679906
- Antigen synonyms:TRPPLIK|CHAK1|LTrpC-7|2310022G15Rik|Ltrpc7|Ltpr7|CHAK|4833414K03Rik|5033407O22Rik
- Storage buffer:0,05% sodium azide
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to C-terminus of rat Trpm7.
- Size:100 μl
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic peptide of Trpm7.
Recommended Dilutions: Immunoprecipitation: 5 ?g/ml lysate; Western Blot: 1:1000
Type: Primary
Antigen: TRPM7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human