Anti-PTPRC Mouse Monoclonal Antibody (Biotin) [clone: MEM-28]
ABNOMAB4578
: Abnova
- Antibody type:Primary
- Antigen name:Protein tyrosine phosphatase receptor type C
- Antigen symbol:PTPRC
- Clonality:Monoclonal
- Clone:MEM-28
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:5788
- Antigen synonyms:CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
- Storage buffer:PBS, pH 7,2 (15 mM sodium azide)
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:Native purified PTPRC from human thymocytes and T lymphocytes.
- Size:100 μg
- Pk:100 µl
Mouse monoclonal antibody raised against native PTPRC.
Recommended Dilutions: Flow Cytometry: 1:500
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: MEM-28
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human