Anti-CD8A Monoclonal Antibody (Biotin) [clone: MEM-31]
Catalog # ABNOMAB4593
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:CD8 antigen, alpha chain
- Antigen symbol:CD8A
- Clonality:Monoclonal
- Clone:MEM-31
- Conjugation:Biotin
- Flow cytometry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:925|926
- Antigen synonyms:p32|Lyt-66|BB154331|Ly-2|Leu2|Ly-35|MAL|CD8|Ly-B
- Storage buffer:PBS, pH 7,2 (15 mM sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified CD8 from Crude thymus membrane fraction.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against native CD8.
Recommended Dilutions: Flow Cytometry: 1:2500
Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: MEM-31
Conjugation: Biotin
Epitope:
Host:
Isotype: IgG2a
Reactivity: Human