Anti-C19ORF57 Rabbit Polyclonal Antibody
ABNOPAB24288
: Abnova
- Antibody type:Primary
- Antigen name:Chromosome 19 open reading frame 57
- Antigen symbol:C19orf57
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:79173
- Storage buffer:PBS, pH 7,5 (40% glycerol, 0,02% sodium azide)
- Sequence:SQIQDALDASDFEAPPEQLFPSGNKPGPCWPGPSSHANGDPVAVAKAQPSRLIMGTHRDLEAFKRLNYRKTKLGGKAPLPYPSKGPGNIPRGDPPWREL
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids of human C19orf57.
- Purification:Antigen affinity purification
- Size:100 μl
- Pk:100 µl
Rabbit polyclonal antibody raised against recombinant C19orf57.
Recommended Dilutions: Immunohistochemistry: 1:200-1:500
Type: Primary
Antigen: C19ORF57
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human