Anti-Quinidine Monoclonal Antibody [clone: 13-406.6.1]
ABNOMAB2250
: Abnova
- Antibody type:Primary
- Antigen name:Quinidine
- Clonality:Monoclonal
- Clone:13-406.6.1
- Conjugation:Unconjugated
- ELISA:Yes
- Isotype:IgG1
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS, pH 7,4
- Storage temperature:Store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Quinidine conjugated with KLH.
- Pk:100 µG
Mouse monoclonal antibody raised against Quinidine.
Type: Primary
Antigen: Quinidine
Clonality: Monoclonal
Clone: 13-406.6.1
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity: