Anti-Lidocaine Monoclonal Antibody [clone: 11-251.3.6]
Catalog # ABNOMAB2254
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Lidocaine
- Clonality:Monoclonal
- Clone:11-251.3.6
- Conjugation:Unconjugated
- ELISA:Yes
- Isotype:IgG1
- Cross adsorption:No
- Format:Liquid
- Form:Liquid
- Storage buffer:In PBS, pH 7,4
- Storage temperature:Store at –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Lidocaine conjugated with KLH.
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against Lidocaine.
Type: Primary
Antigen: Lidocaine
Clonality: Monoclonal
Clone: 11-251.3.6
Conjugation:
Epitope:
Host:
Isotype: IgG1
Reactivity: