Anti-CACH2C Mouse Monoclonal Antibody [clone: S57-46]
ABNOMAB6647
: Abnova
- Antibody type:Primary
- Antigen symbol:CACH2C
- Clonality:Monoclonal
- Clone:S57-46
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Rabbit,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:100144322
- Storage buffer:PBS, pH 7,4 (50% glycerol, 0,09% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant fusion protein corresponding to intracellular C-terminus amino acids 1507-1733 of rabbit CACH2C.
- Size:100 μg
- Pk:100 µl
Mouse monoclonal antibody raised against partial recombinant CACH2C.
Recommended Dilutions: Western Blot: 1-10 ?g/ml; Immunohistochemistry: 0.1-1.0 ?g/ml; Immunocytochemistry: 0.1-1.0 ?g/ml; Immunofluorescence: 1.0-10 ?g/ml
Type: Primary
Antigen: CACH2C
Clonality: Monoclonal
Clone: S57-46
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rabbit, Rat