Anti-HCN1 Mouse Monoclonal Antibody [clone: S70-28]
Catalog # ABNOMAB6651
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:hyperpolarization-activated cyclic nucleotide-gated potassium channel 1
- Antigen symbol:HCN1
- Clonality:Monoclonal
- Clone:S70-28
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:84390
- Storage buffer:PBS, pH 7,4 (50% glycerol, 0,09% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant fusion protein corresponding to C-terminus amino acids 778-910 of rat Hcn1.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant Hcn1.
Recommended Dilutions: Western Blot: 1-10 ?g/ml; Immunohistochemistry: 0.1-1.0 ?g/ml; Immunocytochemistry: 0.1-1.0 ?g/ml; Immunofluorescence: 1.0-10 ?g/ml
Type: Primary
Antigen: HCN1
Clonality: Monoclonal
Clone: S70-28
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat