Order Entry
Export
Orders LinkContactUsLinkComponent
Anti-Eap Rabbit Polyclonal Antibody

Anti-Eap Rabbit Polyclonal Antibody

100 μg
Catalog # ABNOPAB16067
Supplier:   Abnova
Anti-Eap Rabbit Polyclonal Antibody
Catalog # ABNOPAB16067
Supplier:  Abnova
Anti-Eap Rabbit Polyclonal Antibody

100 μg

Price
648,00 €
Log in for availability
1 * 100 µG
Product Details & Documents

Rabbit polyclonal antibody raised against synthetic peptide of Staphylococcus aureus Eap.

Recommended Dilutions: ELISA: 1:2000-1:5000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: Eap
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Staphylococcus

Documents

Specifications
  • Size:
    100 μg
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Antibody type:
    Primary
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Host:
    Rabbit
  • Cross adsorption:
    No
  • Antigen name:
    Staphylococcus aureus Eap
  • Cat. No.:
    ABNOPAB16067
  • Reactivity:
    Staphylococcus
  • Format:
    Liquid
  • Immunogen:
    A synthetic peptide corresponding to C-terminus of Staphylococcus aureus Eap.
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    Eap
  • Storage buffer:
    PBS, pH 7,2 (50% glycerol, 0,01% sodium azide)
  • ELISA:
    Yes