Anti-ABCC12 Rat Monoclonal Antibody [clone: M9I-38]
Catalog # ABNOMAB6674
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:ATP-binding Cassette, Subfamily C (CFTR/MRP), Member 12
- Antigen symbol:ABCC12
- Clonality:Monoclonal
- Clone:M9I-38
- Conjugation:Unconjugated
- Host:Rat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Form:Culture Supernatant
- Gene ID:94160
- Antigen synonyms:MRP9
- Amino acid number:1 to 42
- Storage buffer:0,7% BSA, 0,1% sodium azide
- Storage temperature:Store at 4 °C.
- Immunogen:Recombinant protein corresponding to amino acids 1-42 of human ABCC12.
- Size:1 mL
- Pk:1 ml
Specifications
About this item
Rat monoclonal antibody raised against partial recombinant ABCC12.
Recommended Dilutions: Immunohistochemistry (Frozen sections): 1:20; Immunocytochemistry: 1:20-1:50 Wstern Blot (1:20-1:50
Type: Primary
Antigen: ABCC12
Clonality: Monoclonal
Clone: M9I-38
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG
Reactivity: Human