Anti-ABCA8 Rabbit Polyclonal Antibody
ABNOPAB24373
: Abnova
- Antibody type:Primary
- Antigen name:ATP-binding Cassette, Subfamily A (ABC1), Member 8
- Antigen symbol:ABCA8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:10351
- Antigen synonyms:ATP-binding cassette sub-family A member 8|KIAA0822
- Storage buffer:PBS, pH 7,5 (40% glycerol, 0,02% sodium azide)
- Sequence:KEVLGLPDEESIKEFTANYPEEIVRVTFTNTYSYHLKFLLGHGMPAKKEHKDHTAHCYETNEDVYCEVSVFWKEGFVA
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids of human ABCA8.
- Purification:Antigen affinity purification
- Size:100 μl
- Pk:100 µl
Rabbit polyclonal antibody raised against recombinant ABCA8.
Recommended Dilutions: Immunohistochemistry: 1:50-1:200
Type: Primary
Antigen: ABCA8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human