Anti-AGTR2 Rabbit Polyclonal Antibody
Catalog # ABNOPAB16829
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Angiotensin II Receptor, Type 2
- Antigen symbol:AGTR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:186
- Antigen synonyms:MRX88|Angiotensin II type-2 receptor|AT2|ATGR2|Type-2 angiotensin II receptor
- Storage buffer:PBS (0,02% sodium azide)
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to internal region 16 amino acids of human AGTR2.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of AGTR2.
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: AGTR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat