Anti-NDUFAF2 Rabbit Polyclonal Antibody
Catalog # ABNOPAB16848
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:NADH dehydrogenase (ubiquinone) complex 1, assembly factor 2
- Antigen symbol:NDUFAF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:91942
- Antigen synonyms:Mimitin|Myc-induced mitochondrial protein|B17.2L|mitochondrial|mimitin|MMTN|Mimitin Antibody|NDUFA12L
- Storage buffer:PBS (0,02% sodium azide)
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to C-terminus 14 amino acids of human NDUFAF2.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of NDUFAF2.
Recommended Dilutions: Western Blot: 1-2 ?g/ml
Type: Primary
Antigen: NDUFAF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human