Anti-H7 Mouse Monoclonal Antibody [clone: 1H11]
ABNOMAB8530
: Abnova
- Antibody type:Primary
- Antigen name:Influenza A virus Haemagglutinin H7
- Antigen symbol:H7
- Clonality:Monoclonal
- Clone:1H11
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Virus
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS, pH 7,4 (0,1% sodium azide)
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:Influenza virus A/FPV (H7N1).
- Size:1 mg
- Pk:1 mg
Mouse monoclonal antibody raised against Influenza A haemagglutinin H7 (H7N1).
Type: Primary
Antigen: H7
Clonality: Monoclonal
Clone: 1H11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Virus