Anti-OCIAD2 Rabbit Polyclonal Antibody
Catalog # ABNOPAB16857
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:OCIA Domain Containing 2
- Antigen symbol:OCIAD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:132299
- Antigen synonyms:Ovarian carcinoma immunoreactive antigen-like protein|OCIA domain-containing protein 2|OCIAD2
- Storage buffer:PBS (0,02% sodium azide)
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to N-terminus 17 amino acids of human OCIAD2.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of OCIAD2.
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: OCIAD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat