Anti-CD14 Mouse Monoclonal Antibody [clone: biG 53]
Catalog # ABNOMAB5660
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:CD14 Molecule
- Antigen symbol:CD14
- Clonality:Monoclonal
- Clone:biG 53
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Mouse
- Cross adsorption:No
- Format:Liquid
- Gene ID:12475
- Antigen synonyms:monocyte differentiation antigen CD14|myeloid cell-specific leucine-rich glycoprotein
- Storage buffer:PBS
- Storage temperature:Store at –20 °C. For long term storage store at –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to mouse Cd14.
- Size:100 μg
- Pk:100 Tests
Specifications
About this item
Mouse monoclonal antibody raised against recombinant Cd14.
Recommended Dilutions: ELISA: 1:20000; Flow Cytometry: 1:20000
Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: biG 53
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse