Anti-LBP Mouse Monoclonal Antibody [clone: biG 48]
Catalog # ABNOMAB5662
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Lipopolysaccharide Binding Protein
- Antigen symbol:LBP
- Clonality:Monoclonal
- Clone:biG 48
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Pig,Rabbit,Horse,Human,Bovine,Dog
- Western blot:Yes
- Cross adsorption:No
- Format:Lyophilized
- Gene ID:3929
- Antigen synonyms:LPS-Binding Protein|BPI Fold Containing Family D|BPIFD2|Lipopolysaccharide-Binding Protein|Member 2
- Storage buffer:PBS
- Storage temperature:Store at –20 °C on dry atmosphere. After reconstitution with deionized water, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human LBP.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against recombinant LBP.
Recommended Dilutions: ELISA: 1:10000
Type: Primary
Antigen: LBP
Clonality: Monoclonal
Clone: biG 48
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Dog, Horse, Human, Pig, Rabbit