Anti-CD63 Mouse Monoclonal Antibody [clone: AHN16.1/46-4-5]
Catalog # ABNOMAB6919
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:CD63 molecule
- Antigen symbol:CD63
- Clonality:Monoclonal
- Clone:AHN16.1/46-4-5
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:967
- Antigen synonyms:OMA81H|ME491|LAMP-3|MLA1|TSPAN30
- Storage buffer:50 mM sodium phosphate buffer, 100 mM potassium Chloride, 150 mM NaCl, pH 7,5 (0,5 mg/mL gentamicsulfate)
- Storage temperature:Store at 4 °C.
- Immunogen:Native purified CD63 from human eosinophils.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against native CD63.
Type: Primary
Antigen: CD63
Clonality: Monoclonal
Clone: AHN16.1/46-4-5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human