Anti-ITGAL Mouse Monoclonal Antibody [clone: 38]
Catalog # ABNOMAB6942
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Integrin Alpha L
- Antigen symbol:ITGAL
- Clonality:Monoclonal
- Clone:38
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:3683
- Antigen synonyms:LFA-1 alpha|antigen CD11A p180|CD11 antigen-like family member A|integrin alpha-L|LFA-1A|lymphocyte function-associated antigen 1 alpha polypeptide
- Storage buffer:50 mM sodium phosphate buffer, 100 mM potassium Chloride, 150 mM NaCl, pH 7,5 (0,5 mg/mL gentamicsulfate)
- Storage temperature:Store at 4 °C.
- Immunogen:Native purified ITGAL from human monocytes.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against native ITGAL.
Type: Primary
Antigen: ITGAL
Clonality: Monoclonal
Clone: 38
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human