Anti-SELL Rat Monoclonal Antibody (Spectral Red®) [clone: MEL-14]
Catalog # ABNOMAB5739
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Selectin L
- Antigen symbol:SELL
- Clonality:Monoclonal
- Clone:MEL-14
- Conjugation:Spectral Red®
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Cross adsorption:No
- Format:Liquid
- Gene ID:20343
- Antigen synonyms:LECAM1|CD62L|TQ1|LYAM1|LEU8|LNHR|LSEL|LAM1|PLNHR
- Storage buffer:PBS (1 mg/mL sodium azide)
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:C3H/eb cloned mouse B lymphoma 38C-13.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rat monoclonal antibody raised against Sell.
Recommended Dilutions: Flow Cytometry: 0.1 ?g/10^6 cells
Type: Primary
Antigen: SELL
Clonality: Monoclonal
Clone: MEL-14
Conjugation: Spectral Red®
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse