Anti-TRPV3 Mouse Monoclonal Antibody [clone: S15-39]
Catalog # ABNOMAB7896
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Transient Receptor Potential Cation Channel Subfamily V member 3
- Antigen symbol:TRPV3
- Clonality:Monoclonal
- Clone:S15-39
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:497948
- Storage buffer:PBS, pH 7,4 (50% glycerol, 0,09% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to C-terminus amino acids 458-474 of rat Kcnt2.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against synthetic peptide of Kcnt2.
Recommended Dilutions: Western Blot: 1-10 ?g/ml; Immunohistochemistry: 0.1-1.0 ?g/ml; Immunocytochemistry: 0.1-1.0 ?g/ml; Immunofluorescence: 1-10 ?g/ml
Type: Primary
Antigen: TRPV3
Clonality: Monoclonal
Clone: S15-39
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat