Anti-SHANK3 Mouse Monoclonal Antibody [clone: S69-46]
Catalog # ABNOMAB7898
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:SH3 and multiple ankyrin repeat domains 3
- Antigen symbol:SHANK3
- Clonality:Monoclonal
- Clone:S69-46
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:59312
- Antigen synonyms:PROSAP2|Shank3|SCZD15|SHANK3|DEL22q13.3|PSAP2|Proline-rich synapse-associated protein 2|KIAA1650|SPANK-2|SH3 and multiple ankyrin repeat domains protein 3
- Amino acid number:840 to 857
- Storage buffer:PBS, pH 7,4 (50% glycerol, 0,09% sodium azide)
- Sequence:PEKLPGSLRKGIPRTKSV
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 840-857 of rat Kcnu1.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against synthetic peptide of Kcnu1.
Recommended Dilutions: Western Blot: 1-10 ?g/ml; Immunohistochemistry: 0.1-1.0 ?g/ml; Immunocytochemistry: 0.1-1.0 ?g/ml; Immunofluorescence: 1-10 ?g/ml
Type: Primary
Antigen: SHANK3
Clonality: Monoclonal
Clone: S69-46
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat