Order Entry
Export
ContactUsLinkComponent
Anti-TRPC7 Mouse Monoclonal Antibody [clone: S64A-36]
Anti-TRPC7 Mouse Monoclonal Antibody [clone: S64A-36]
Catalog # ABNOMAB7904
Supplier:  Abnova
Anti-TRPC7 Mouse Monoclonal Antibody [clone: S64A-36]
Catalog # ABNOMAB7904
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Transient Receptor Potential Cation Channel Subfamily C member 7)
  • Antigen symbol:
    TRPC7
  • Clonality:
    Monoclonal
  • Clone:
    S64A-36
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    57113
  • Antigen synonyms:
    TRP7 protein|mTRP7|Trrp8|TRP7|Trpc7|TrpC7|Trp7|Short transient receptor potential channel 7
  • Amino acid number:
    845 to 862
  • Storage buffer:
    PBS, pH 7,4 (50% glycerol, 0,09% sodium azide)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to amino acids 845-862 of human Cacnb2.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against synthetic peptide of Cacnb2.

Recommended Dilutions: Western Blot: 1-10 ?g/ml; Immunohistochemistry: 0.1-1.0 ?g/ml; Immunocytochemistry: 0.1-1.0 ?g/ml; Immunofluorescence: 1-10 ?g/ml

Type: Primary
Antigen: TRPC7
Clonality: Monoclonal
Clone: S64A-36
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat