Anti-TNFRSF8 Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: MEM-268]
Catalog # ABNOMAB5099
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Tumor Necrosis Factor Receptor Superfamily Member 8
- Antigen symbol:TNFRSF8
- Clonality:Monoclonal
- Clone:MEM-268
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:943
- Antigen synonyms:lymphocyte activation antigen CD30|CD30|cytokine receptor CD30|tumor necrosis factor receptor superfamily member 8|Ki-1 antigen|CD30L receptor|CD30 antigen
- Storage buffer:PBS (0,2% BSA, 15 mM sodium azide)
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Expression vector containing CD30 cDNA (booster suspension of THP-1 cell line).
- Size:100 Reactions
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against TNFRSF8.
Recommended Dilutions: Flow Cytometry: 20 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)
Type: Primary
Antigen: TNFRSF8
Clonality: Monoclonal
Clone: MEM-268
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host:
Isotype: IgG
Reactivity: Human