Order Entry
Export
ContactUsLinkComponent
Anti-TNFRSF10D Monoclonal Antibody (PE (Phycoerythrin)) [clone: TRAIL-R4-01]
Anti-TNFRSF10D Monoclonal Antibody (PE (Phycoerythrin)) [clone: TRAIL-R4-01]
  ABNOMAB5140
 :  Abnova
Anti-TNFRSF10D Monoclonal Antibody (PE (Phycoerythrin)) [clone: TRAIL-R4-01]
  ABNOMAB5140
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Tumor Necrosis Factor Receptor Superfamily Member 10D
  • Antigen symbol:
    TNFRSF10D
  • Clonality:
    Monoclonal
  • Clone:
    TRAIL-R4-01
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    8793
  • Antigen synonyms:
    TRAILR4|TRAIL-R4|CD264|TRUNDD|DCR2
  • Amino acid number:
    1 to 210
  • Storage buffer:
    PBS (0,2% BSA, 15 mM sodium azide)
  • Storage temperature:
    Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant Fc fusion protein corresponding to amino acids 1-210 of TNFRSF10D.
  • Size:
    100 μg
  • Pk:
    100 µg

 

 

Mouse monoclonal antibody raised against partial recombinant TNFRSF10D.

Recommended Dilutions: Flow Cytometry: 3 ?g/ml

Type: Primary
Antigen: TNFRSF10D
Clonality: Monoclonal
Clone: TRAIL-R4-01
Conjugation: PE (Phycoerythrin)
Epitope:
Host:
Isotype: IgG1
Reactivity: Human