Anti-Clostridium botulinum Type A Hemagglutinin 33 Rabbit Polyclonal Antibody
Catalog # ABNOPAB13911
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Clostridium botulinum Type A Hemagglutinin 33
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS, pH 7,2 (20% glycerol)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Clostridium botulinum type A Hemagglutinin 33.
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Rabbit polyclonal antibody raised against Clostridium botulinum Type A Hemagglutinin 33.
Recommended Dilutions: ELISA: 1:10000
Type: Primary
Antigen: Clostridium botulinum Type A Hemagglutinin 33
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: