Anti-SAA Mouse Monoclonal Antibody [clone: 38]
Catalog # ABNOMAB6414
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Serum Amyloid A
- Antigen symbol:SAA
- Clonality:Monoclonal
- Clone:38
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Lyophilized
- Antigen synonyms:PIG4|serum amyloid A protein|SAA|tumor protein p53 inducible protein 4|TP53I4|MGC111216
- Storage buffer:10 mM PBS, pH 7,2
- Storage temperature:Store at –20 °C on dry atmosphere. After reconstitution with deionized water, store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human Serum Amyloid A (SAA).
- Size:500 μg
- Pk:500 µg
Specifications
About this item
Mouse monoclonal antibody raised against recombinant SAA.
Recommended Dilutions: Western Blot: 0.1-0.5 ?g
Type: Primary
Antigen: SAA
Clonality: Monoclonal
Clone: 38
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human