Anti-ITGAX Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 3.9]
Catalog # ABNOMAB6037
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Integrin Alpha 10
- Antigen symbol:ITGA10
- Clonality:Monoclonal
- Clone:3.9
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:3687
- Antigen synonyms:integrin alpha-X|integrin aX|ITGAX|AI449405|CD11 antigen-like family member C|N418|Cr4|Cd11c
- Storage buffer:PBS (0,1 mg/mL sodium azide)
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified from human rheumatoid synovial fluid cells and fibronectin purified human monocytes.
- Size:100 Reactions
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against native ITGAX.
Recommended Dilutions: Flow Cytometry: 20 ?l/10^6 cells
Type: Primary
Antigen: ITGAX
Clonality: Monoclonal
Clone: 3.9
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human