Anti-FUT4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 28]
Catalog # ABNOMAB6049
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Fucosyltransferase 4
- Antigen symbol:FUT4
- Clonality:Monoclonal
- Clone:28
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgM
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:2526
- Antigen synonyms:CD15|FUC-TIV|ELFT|SSEA-1|LeX|FCT3A|FUTIV
- Storage buffer:PBS (0,1 mg/mL sodium azide)
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified from human monocytes, separated from other peripheral blood leucocytes on fibronectin plates.
- Size:100 Reactions
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against native FUT4.
Recommended Dilutions: Flow Cytometry: 10 ?l/10^6 cells
Type: Primary
Antigen: FUT4
Clonality: Monoclonal
Clone: 28
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human