Anti-HLAA Mouse Monoclonal Antibody [clone: W6/32]
Catalog # ABNOMAB6449
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:major histocompatibility complex, class I, A
- Antigen symbol:HLAA
- Clonality:Monoclonal
- Clone:W6/32
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Primates,Human,Bovine,Cat
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:3105
- Antigen synonyms:HLAA
- Storage buffer:PBS, pH 7,4
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:Native purified HLA-A from membrane of human tonsil cells.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against native HLA-A.
Type: Primary
Antigen: HLAA
Clonality: Monoclonal
Clone: W6/32
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Bovine, Cat, Human, Non-Human Primates