Anti-RNF39 Goat Polyclonal Antibody
Catalog # ABNOPAB6326
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ring Finger Protein 39
- Antigen symbol:RNF39
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:80352
- Antigen synonyms:RNF39|HZFW1|LTP (long-term potentiation) induced RING finger protein|LIRF|HZF|HZFW|DADB-60O11.8|HZFw1 protein
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:CDPRAPLRIVPAES
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human RNF39.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of RNF39.
Recommended Dilutions: ELISA: 1:16000; Western Blot: 1-3 ?g/ml
Type: Primary
Antigen: RNF39
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human