Anti-DNMT3B Rabbit Polyclonal Antibody
ABNOPAB14046
: Abnova
- Antibody type:Primary
- Antigen name:DNA (cytosine-5-)-methyltransferase 3 beta
- Antigen symbol:DNMT3B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Form:Serum
- Gene ID:13436
- Antigen synonyms:ICF|DNA MTase HsaIIIB|DNA (cytosine-5)-methyltransferase 3B|ICF1|RP5-1085F17.2|DNA methyltransferase HsaIIIB|M.HsaIIIB
- Storage buffer:0,05% sodium azide
- Storage temperature:Store at –20 °C. For long term storage store at –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide (conjugated with KLH) corresponding to N-terminus of mouse Dnmt3b.
- Size:100 μl
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic peptide of Dnmt3b.
Recommended Dilutions: ELISA: 1:100-1:500; Western Blot: 1:1000
Type: Primary
Antigen: DNMT3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse